Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Halcinonide | 3093-35-4 | 98.3% | 200 MG
SDP

Catalog No. 5000408194
Encompass_Preferred

Halcinonide is a high potency corticosteroid used topically in the treatment of certain skin conditions. This product is for research use only and not sold to patients.

  • High potency corticosteroid
  • Used topically for skin conditions
  • For research use only
  • Solid appearance
  • Light yellow to yellow color
  • Molecular weight: 454.96
  • Formula: C24H32ClFO5

Catalog No. 50-004-08194 Supplier Medchemexpress LLC Supplier No. HYB0877200MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Halcinonide is a high potency corticosteroid used topically in the treatment of certain skin conditions. This product is for research use only and not sold to patients.

  • High potency corticosteroid
  • Used topically for skin conditions
  • For research use only
  • Solid appearance
  • Light yellow to yellow color
  • Molecular weight: 454.96
  • Formula: C24H32ClFO5

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.