Learn More
Hach Company Half Cable 25 ft- 6 Pin
Supplier:  Hach Company 541
Encompass
Multi-purpose half cable 25ft- 6 Pin Auxiliary connector on one end open leads on the other end Connects a 900Max sampler to a non-Sigma flow memter
Catalog No. 50-002-57167