Learn More
Hach Company Half Cable for Hach SD900 Sampler, 7-Pin, 25 ft (open leads)
Catalog No. 5000207178 Shop All Hach Company Products
Encompass
Half Cable for SD900, 25 ft, is a multi-purpose half cable designed for use with SD900 samplers. It features a 7-pin connector on one end and open leads on the other, providing a straightforward way to connect the sampler to external wiring or devices in field sampling setups.
  • 25 ft (7.62 m) length for extended reach.
  • 7-pin connector on one end for direct instrument connection.
  • Open leads on the other end for easy wiring to external devices.
  • Multi-purpose design suitable as a replacement or accessory cable.
Catalog No. 50-002-07178 Supplier Hach Company Supplier No. 8757200
Half Cable for SD900, 25 ft, is a multi-purpose half cable designed for use with SD900 samplers. It features a 7-pin connector on one end and open leads on the other, providing a straightforward way to connect the sampler to external wiring or devices in field sampling setups.
  • 25 ft (7.62 m) length for extended reach.
  • 7-pin connector on one end for direct instrument connection.
  • Open leads on the other end for easy wiring to external devices.
  • Multi-purpose design suitable as a replacement or accessory cable.