Learn More
MSC HALF FACE PC REUSE RESP
Catalog No. 5000802203
Encompass
This half facepiece respirator, made of silicone with a bayonet connection, provides comfort and durability through its resilient faceseal. A proprietary Cool Flow valve reduces heat and moisture, and an exhalation valve cover directs breath downward, preventing eyewear fogging. Its low-profile design offers a wider field of view and improved compatibility with welding/grinding shields.
  • Resilient silicone faceseal for comfort and durability
  • Proprietary Cool Flow valve reduces heat and moisture
  • Exhalation valve cover directs breath downward to reduce eyewear fogging
  • Low-profile design offers wide field of view and compatibility with shields
  • Adjustable head harness for improved fit and comfort
  • Facepiece type: half mask
  • Connection type: bayonet
  • Size: medium
  • Material: silicone
  • Suspension: 4-point
Catalog No. 50-008-02203 Supplier MSC Supplier No. 68956879
This half facepiece respirator, made of silicone with a bayonet connection, provides comfort and durability through its resilient faceseal. A proprietary Cool Flow valve reduces heat and moisture, and an exhalation valve cover directs breath downward, preventing eyewear fogging. Its low-profile design offers a wider field of view and improved compatibility with welding/grinding shields.
  • Resilient silicone faceseal for comfort and durability
  • Proprietary Cool Flow valve reduces heat and moisture
  • Exhalation valve cover directs breath downward to reduce eyewear fogging
  • Low-profile design offers wide field of view and compatibility with shields
  • Adjustable head harness for improved fit and comfort
  • Facepiece type: half mask
  • Connection type: bayonet
  • Size: medium
  • Material: silicone
  • Suspension: 4-point