Learn More
MSC HALF FACE PC REUSE RESP
Catalog No. 5000766613
Encompass
This Half Facepiece Respirator is constructed from silicone with a bayonet connection, designed for comfort and ease of use in various work environments. It is part of the 6500 Series.
  • Resilient silicone faceseal provides comfort, durability, and stability
  • Quick Latch mechanism allows the respirator to be dropped down from the face without removing head straps
  • Cool Flow valve helps reduce heat and moisture for easier breathing
  • Low-profile design offers a wide field of view and compatibility with welding and grinding shields
  • Exhalation valve cover directs exhaled breath and moisture downward to reduce fogging of face shields
  • Features a 4-point suspension system
Catalog No. 50-007-66613 Supplier MSC Supplier No. 68956853
This Half Facepiece Respirator is constructed from silicone with a bayonet connection, designed for comfort and ease of use in various work environments. It is part of the 6500 Series.
  • Resilient silicone faceseal provides comfort, durability, and stability
  • Quick Latch mechanism allows the respirator to be dropped down from the face without removing head straps
  • Cool Flow valve helps reduce heat and moisture for easier breathing
  • Low-profile design offers a wide field of view and compatibility with welding and grinding shields
  • Exhalation valve cover directs exhaled breath and moisture downward to reduce fogging of face shields
  • Features a 4-point suspension system