Learn More
MSC HALF FACE PC REUSE RESP
Catalog No. 5000803543
Encompass
This half facepiece reusable respirator is designed for comfort, durability, and stability in tough work environments. It features a quick latch mechanism for easy donning and doffing without removing head protection, and a resilient silicone faceseal for a soft yet firm seal.
  • Quick latch drop-down mechanism allows easy on and off
  • Durable design for demanding work environments
  • Resilient silicone faceseal provides comfort and stability
  • Cool Flow valve helps reduce heat and moisture
  • Low-profile design offers a wide field of view
  • Exhalation valve cover directs breath downward
  • Compatible with bayonet-style filters and cartridges
  • Adjustable head harness for a personalized fit
  • NIOSH-approved for respiratory protection
Catalog No. 50-008-03543 Supplier MSC Supplier No. 68956846
This half facepiece reusable respirator is designed for comfort, durability, and stability in tough work environments. It features a quick latch mechanism for easy donning and doffing without removing head protection, and a resilient silicone faceseal for a soft yet firm seal.
  • Quick latch drop-down mechanism allows easy on and off
  • Durable design for demanding work environments
  • Resilient silicone faceseal provides comfort and stability
  • Cool Flow valve helps reduce heat and moisture
  • Low-profile design offers a wide field of view
  • Exhalation valve cover directs breath downward
  • Compatible with bayonet-style filters and cartridges
  • Adjustable head harness for a personalized fit
  • NIOSH-approved for respiratory protection