Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ Half-Finger Medical Nylon Glove Liners

Catalog No. 19044156 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Gender:
Men's
Women's
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Gender
19-044156 Women's
19-044157 Men's
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-044156 Supplier Wells Lamont™ Supplier No. M117W
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Half-Finger Medical Nylon Glove Liners are constructed of high-quality, 100% continuous filament nylon and packaged in a secure box. Wear under a disposable glove for additional comfort, or to reduce hand fatigue from perspiration.

  • 100% continuous nylon filament creates an ideal barrier between hands and disposable gloves
  • Reduces allergies and adverse reactions that can be caused by prolonged exposure to latex
  • Packaged in a secure box so that liners remain clean
  • Fully launderable up to 20 times without shrinking
  • Half finger, regular cuff design
  • Non-sterile
  • Made in US

Specifications

Applications Healthcare
Coating Material Uncoated
Color White
Disposable Reusable
Hand Ambidextrous
Latex Free Yes
Material Nylon
Product Type Medical Glove Liner
Sterility Non-sterile
Finger Style Half
For Use With (Application) Reducing Latex Allerigies and Adverse Reactions
Gender Women's
Glove Cuff Style Knit
Packaging Type Box
Series M117
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.