Learn More
NEOGEN Half Fraser (Demi-Fraser) Broth (ISO), 10 kg
SDP
Catalog No. 502015158
Encompass_Preferred
Half Fraser (Demi-Fraser) Broth is used with ferric ammonium citrate for the selective enrichment of Listeria species and is not intended for use in the diagnosis of disease or other conditions in humans. Primary enrichment broth for the isolation of Listeria spp. formulated according to ISO 11290. The selective components acriflavine and nalidixic acid are blended into the base powder and the ferric ammonium citrate is added to the tempered broth after sterilization. Half Fraser (Demi-Fraser) Broth, NCM0001 was developed to give improved results for the isolation of Listeria spp. with both traditional and ELISA methods. As with the sister product, Fraser Broth, NCM0050 the selective components acriflavine and nalidixic acid are blended into the base powder and the Ferric Ammonium Citrate is added to the tempered broth after autoclaving.For more information visit https://www.neogen.com/solutions/Microbiology/half-fraser-broth
Catalog No. 50-201-5158 Supplier NEOGEN Supplier No. 700002947
Half Fraser (Demi-Fraser) Broth is used with ferric ammonium citrate for the selective enrichment of Listeria species and is not intended for use in the diagnosis of disease or other conditions in humans. Primary enrichment broth for the isolation of Listeria spp. formulated according to ISO 11290. The selective components acriflavine and nalidixic acid are blended into the base powder and the ferric ammonium citrate is added to the tempered broth after sterilization. Half Fraser (Demi-Fraser) Broth, NCM0001 was developed to give improved results for the isolation of Listeria spp. with both traditional and ELISA methods. As with the sister product, Fraser Broth, NCM0050 the selective components acriflavine and nalidixic acid are blended into the base powder and the Ferric Ammonium Citrate is added to the tempered broth after autoclaving.For more information visit https://www.neogen.com/solutions/Microbiology/half-fraser-broth