Learn More
Miele, Inc. E 106 Half Insert with spring clips (18 clips 105 mm tall, 10 clips 175 mm tall)
Catalog No. NC0545150
Encompass_Preferred
E 106 Mesh Basket
For the optimum loading of wide neck flasks, measuring cylinders, etc.
Equipped with 10 spring hooks x 175 mm height & 16 x 105 mm height
The distance between the spring hooks is approx. 60 mm
Visit www.mieleusa.com/professional for technical information.
Catalog No. NC0545150 Supplier Miele, Inc. Supplier No. 03808310
E 106 Mesh Basket
For the optimum loading of wide neck flasks, measuring cylinders, etc.
Equipped with 10 spring hooks x 175 mm height & 16 x 105 mm height
The distance between the spring hooks is approx. 60 mm
Visit www.mieleusa.com/professional for technical information.