Learn More
Miele, Inc. E 109 Half Insert for beakers (21 beakers, up to 250 ml)
Catalog No. NC0545146
Encompass_Preferred
E 109 Mesh Basket
For the optimum loading of beakers up to 250 ml.
Suitable for 21 glass beakers up to 250 ml
Equipped with 21 x 3 stay bars
For use in upper and lower basket
Visit www.mieleusa.com/professional for technical information.
Catalog No. NC0545146 Supplier Miele, Inc. Supplier No. 03808360
E 109 Mesh Basket
For the optimum loading of beakers up to 250 ml.
Suitable for 21 glass beakers up to 250 ml
Equipped with 21 x 3 stay bars
For use in upper and lower basket
Visit www.mieleusa.com/professional for technical information.