Learn More
CleanSpace™ Half Mask Adaptor
Catalog No. 17800621 Shop All Cleanspace Technology Products
Change view
Click to view available options
For Use With (Equipment):
PAF-1010, PAF-0027, PAF-0033
Catalog No. For Use With (Equipment)
17-800-621 PAF-1010, PAF-0027, PAF-0033
1 options
This item is not returnable. View return policy
Catalog No. 17-800-621 Supplier CleanSpace™ Supplier No. PAF0025
Add to Cart
Add to Cart
This item is not returnable. View return policy
Half mask adaptor is equipped for the front of the mask to enable quantitative fit testing.
  • Used with the revolutionary CleanSpace™ Respirators: light weight, no hoses or belts
  • Used with the CleanSpace™ Half Masks only
  • Quick and easy to install
  • Fits with the TSI PortaCount equipment
  • Easy to wash and quick drying
Specifications
Product Type Half Mask Adapter
For Use With (Application) Quantitative Fit Testing
For Use With (Equipment) PAF-1010, PAF-0027, PAF-0033
Model PAF-0025