Learn More
MSC HALF MASK FACE RESPIRATOR
Catalog No. 5000788872
Encompass
The 3M Secure Click Half Facepiece Reusable Respirator, HF-800 Series, is designed to provide respiratory protection against particulates and a wide variety of gases and vapors. It integrates innovative features for enhanced user confidence and comfort, including a secure connection system and an easy seal check.
  • Speaking diaphragm for clear communication
  • Secure Click connection ensures proper cartridge and filter installation
  • Push-button seal check for pre-use verification
  • Silicone flex-joint faceseal for enhanced comfort
  • Easy-adjust buckle for simple two-way adjustability
  • Dual-flow cartridge design for easy breathing
  • Low-profile design for wide field of view and shield compatibility
  • NIOSH-approved for respiratory protection
Catalog No. 50-007-88872 Supplier MSC Supplier No. 18772863
The 3M Secure Click Half Facepiece Reusable Respirator, HF-800 Series, is designed to provide respiratory protection against particulates and a wide variety of gases and vapors. It integrates innovative features for enhanced user confidence and comfort, including a secure connection system and an easy seal check.
  • Speaking diaphragm for clear communication
  • Secure Click connection ensures proper cartridge and filter installation
  • Push-button seal check for pre-use verification
  • Silicone flex-joint faceseal for enhanced comfort
  • Easy-adjust buckle for simple two-way adjustability
  • Dual-flow cartridge design for easy breathing
  • Low-profile design for wide field of view and shield compatibility
  • NIOSH-approved for respiratory protection