Learn More
Ampco Half Round File
SDP
Catalog No. 19173522 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
Length (English):
14 -1/2 in.
Catalog No. Length (English)
19-173-522 14 -1/2 in.
1 options
Catalog No. 19-173-522 Supplier Ampco Supplier No. 8380
Add to Cart
Add to Cart
Non-sparking, non-magnetic, and corrosion-resistant half roundfile.
  • Please see image for additional information regarding measurements.
  • Tip size (a): 1 in.
  • Additional tip measurement (b): 3/8 in.,
  • File length (c): 10 and 1/4 in.
  • Overall length: 14 and 1/2 in.
Specifications
Product Type Half Round File
Length (English) 14 -1/2 in.
Length (English) Blade 10 -1/4 in.
Quantity 1 Ea.