Learn More
MOLTOX Molecular Toxicology Inc Half Strength Nutrient Agar, 20 mL/plate, 20 plates/sleeve, Store in Refrigerator
SDP
Catalog No. 50948703
Encompass_Preferred
Half Strength Nutrient Agar is commonly used for the identification and enumeration of total bacteria and Actinomyces, whereas the low nutrient content reduces the overgrowth of unwanted microorganisms for optimal enumeration of the species of interest. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-703 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 2140S95
Half Strength Nutrient Agar is commonly used for the identification and enumeration of total bacteria and Actinomyces, whereas the low nutrient content reduces the overgrowth of unwanted microorganisms for optimal enumeration of the species of interest. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.