Learn More
Medicus Health Half Tube Rack For 16Mm Tubes, Green, 5"L x 5"W x 2.75"H
SDP
Catalog No. NC1244052
Encompass
Half Tube Racks, Ultra Durable Delrin for 16mm Tubes Holds up to thirty six tubes 16mm in diameter ∙Tube Rack made from Delrin designed for the toughest conditions∙Constructed of Delrin material so can handle extreme cold and hot temperatures (-40 F - 250 F)∙Resistant to acids, solvents, and most chemicals∙Rack has 36 places for up to 16mm diameter tubes∙Fits in Droplet Phlebotomy Tray 5252M2∙Available in four colors Red, White, Blue and Green - Please specify color when ordering∙Mix and match any color and quantity∙Includes molded-in index coordinates and flat areas for labeling∙Autoclavable∙Dimensions: 5"L x 5"W x 2.75"H
Catalog No. NC1244052 Supplier Medicus Health Supplier No. 5257M1GREEN
Add to Cart
Add to Cart
Half Tube Racks, Ultra Durable Delrin for 16mm Tubes Holds up to thirty six tubes 16mm in diameter ∙Tube Rack made from Delrin designed for the toughest conditions∙Constructed of Delrin material so can handle extreme cold and hot temperatures (-40 F - 250 F)∙Resistant to acids, solvents, and most chemicals∙Rack has 36 places for up to 16mm diameter tubes∙Fits in Droplet Phlebotomy Tray 5252M2∙Available in four colors Red, White, Blue and Green - Please specify color when ordering∙Mix and match any color and quantity∙Includes molded-in index coordinates and flat areas for labeling∙Autoclavable∙Dimensions: 5"L x 5"W x 2.75"H