Learn More
Cayman Chemical Half Volume 96Well Solid Plat
SDP
Catalog No. 502886616
Encompass_Preferred
An uncoated 96-well plate for use in our SIRT1 FRET-Based Screening Assay Kit (Catalog No. 10010991) and SIRT1 Direct Fluorescent Screening Assay Kit (Catalog No.
Catalog No. 50-288-6616 Supplier Cayman Chemical Supplier No. 100112881 EA
Add to Cart
Add to Cart
An uncoated 96-well plate for use in our SIRT1 FRET-Based Screening Assay Kit (Catalog No. 10010991) and SIRT1 Direct Fluorescent Screening Assay Kit (Catalog No.