Learn More
FUJIFILM Biosciences Halfenprox Standard
Catalog No. 5000450351
Encompass_Preferred
Highly pure analytical standard 98% used for pesticide residue testing Store at -20 C under inert gas
Catalog No. 50-004-50351 Supplier FUJIFILM Biosciences Supplier No. 08807101
Highly pure analytical standard 98% used for pesticide residue testing Store at -20 C under inert gas