Learn More
Braintree Scientific Hallowell Intubation Pack for Rats
SDP
Supplier:  Braintree Scientific RWA3746
Encompass
Hallowell Intubation Pack - For use in conjunction with the Rodent WorkStand. The intubation packs, a rat pack and a mouse pack, hold everything needed to directly visualize and quickly intubate with a minimum amount of trauma. Each pack is designed to hold all tools and supplies needed to perform quick, efficient and minimally traumatic orotracheal intubation of small rodents. Intubation packs include specially designed and molded, autoclavable intubation speculum, lidocaine applicator, endotracheal tube guide wire, incisor loop and a brief video tutorial of how to perform intubations. Rat Pack also contains scissors, umbilical tape, and mirror for the verification of tracheal intubation. Mouse pack includes two sizes of Hallowell's mouse endotracheal tubes. Packaged neatly and conveniently in a 13" x 8" x 2" polypropylene organizer box with each compartment labeled for contents and reordering information. Can be stored most anywhere and always ready for use.
Catalog No. 50-195-5815
 Restricted Item
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov