Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Halloysite nanoclay

Catalog No. 685445500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
68-544-5500G 500 g
68-544-5100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 68-544-5500G Supplier Sigma Aldrich Supplier No. 685445500G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Halloysite nanoclay has a tube like morphology. It shows low electrical, thermal conductivity and strong hydrogen interactions, on account of which the inner hydroxyl groups show greater stability than the surface hydroxyl groups in halloysite. The loss of hole water occurs at nearly 300°C.Similar to montmorillonite, halloysite consists of two layers of aluminosilicate in 1:1 Al:Si ratio. Significant improvements in thermal, mechanical properties have been reported on addition of halloysite nanotubes in polymer composites.Halloysite is frequently used as a reinforcement in various nanocomposites, some of which are listed here:Isotactic propylene-halloysite nanocomposites.Alloy composites by electroless/ autocatalytic deposition. Poly(butylene terephthalate) (PBT) composites.Zinc-halloysite nanotube (HNT) composite coatings.Halloysite nanotube-hydroxyapatite-hyaluronic acid films.Halloysite combined with polyamide, polypropylene, poly(α-caprolactone) and fluroelastomers.Its tube like morphology, high aspect ratio, and low percolation enables interesting applications as reinforcements in nanocomposites.Product of Applied Minerals, Inc. Customers may obtain additional technical support and purchase commercial quantities at appliedminerals.com

Specifications

CAS 1332-58-7
Density 2.53 (true specific gravity)
Molecular Formula H4Al2O9Si2 -+ 2 H2O
Linear Formula Al2Si2O5(OH)4 -+ 2 H2O
Refractive Index n20/D 1.54
MDL Number MFCD11045913
Quantity 500 g
Synonym Kaolin clay
Molecular Weight (g/mol) 294.19
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.