Learn More
Med Vet International Halls Anesthesia Mask Medium, J0298HB
Catalog No. 5000582335
Encompass_Preferred
Soft, malleable black rubber. This mask will conform to the animal's muzzle shape, allowing a custom fit. Standard 15mm connection to anesthesia circuit.
Catalog No. 50-005-82335 Supplier Med Vet International Supplier No. JOR298HB
Soft, malleable black rubber. This mask will conform to the animal's muzzle shape, allowing a custom fit. Standard 15mm connection to anesthesia circuit.