Learn More
Med Vet International Halls Anesthesia Mask Small, J0298HA
Catalog No. 5000583139
Encompass_Preferred
Soft, malleable black rubber. This mask will conform to the animal's muzzle shape, allowing a custom fit. Standard 15mm connection to anesthesia circuit.
Catalog No. 50-005-83139 Supplier Med Vet International Supplier No. JOR298HA
Soft, malleable black rubber. This mask will conform to the animal's muzzle shape, allowing a custom fit. Standard 15mm connection to anesthesia circuit.