Learn More
Advanced Materials Technology HALO 1000 C4 2.7 3.0 X
Supplier:  Advanced Materials Technology 92713614
Encompass_Preferred
HALO BIOCLASS C4 1000 Å Protein columns made with innovative large pore Fused-Core particle technology provide best in class performance for high resolution separations and better recoveries of large biomolecules such as mAbs, versus smaller pore size columns. HALO 1000 Å columns are optimized for high performance in HPLC, UHPLC, and LCMS large molecule applications delivering excellent reproducibility and column lifetimes.
Catalog No. 50-255-6407