Learn More
Advanced Materials Technology HALO 1000 ES-C18 2.7 0.
Catalog No. 502555653
Encompass_Preferred
HALO BIOCLASS ES-C18 1000 Å Protein columns made with innovative large pore Fused-Core particle technology provide best in class performance for high resolution separations and better recoveries of large biomolecules such as mAbs, versus smaller pore size columns. HALO 1000 Å columns are optimized for high performance in HPLC, UHPLC, and LCMS large molecule applications delivering excellent reproducibility and column lifetimes.
Catalog No. 50-255-5653 Supplier Advanced Materials Technology Supplier No. 97215402
Add to Cart
Add to Cart
HALO BIOCLASS ES-C18 1000 Å Protein columns made with innovative large pore Fused-Core particle technology provide best in class performance for high resolution separations and better recoveries of large biomolecules such as mAbs, versus smaller pore size columns. HALO 1000 Å columns are optimized for high performance in HPLC, UHPLC, and LCMS large molecule applications delivering excellent reproducibility and column lifetimes.