Learn More
Advanced Materials Technology HALO 1000 Å OLIGO C18, 2.7 µm, 2.1 mm x 30 mm - INERT
Catalog No. 5000332634
Encompass_Preferred
HALO BIOCLASS Oligonucleotide columns are designed with the foundational properties of Fused-Core technology for high efficiency, high speed separations and optimized for the needs of oligonucleotide separations. The HALO 1000 Å OLIGO C18 columns deliver superior separation and impurity profiling for longchain oligonucleotides, (up to and beyond 100 nucleotides) with higher peak capacity and lower backpressure. Ideal for advanced applications like CRISPR and therapeutic development. Their robust design supports high pH and temperature conditions, scalable column lengths, andreliable performance for advanced applications The inert column hardware also addresses absorptivity concerns so the highest recoveries can be achieved.
Catalog No. 50-003-32634 Supplier Advanced Materials Technology Supplier No. P2762302
HALO BIOCLASS Oligonucleotide columns are designed with the foundational properties of Fused-Core technology for high efficiency, high speed separations and optimized for the needs of oligonucleotide separations. The HALO 1000 Å OLIGO C18 columns deliver superior separation and impurity profiling for longchain oligonucleotides, (up to and beyond 100 nucleotides) with higher peak capacity and lower backpressure. Ideal for advanced applications like CRISPR and therapeutic development. Their robust design supports high pH and temperature conditions, scalable column lengths, andreliable performance for advanced applications The inert column hardware also addresses absorptivity concerns so the highest recoveries can be achieved.