Learn More
Advanced Materials Technology HALO 120 A OLIGO C18 2.7 um 4.6 x 50 mm
Catalog No. NC3596289
Encompass_Preferred
The HALO OLIGO C18 incorporates surface modified organo-silane technology for alkaline resistance, resulting in excellent stability under elevated pH operating conditions common in oligonucleotide separation methods.  The inert column hardware also addresses absorptivity concerns so the highest recoveries can be achieved.
Catalog No. NC3596289 Supplier Advanced Materials Technology Supplier No. P2A64402
The HALO OLIGO C18 incorporates surface modified organo-silane technology for alkaline resistance, resulting in excellent stability under elevated pH operating conditions common in oligonucleotide separation methods.  The inert column hardware also addresses absorptivity concerns so the highest recoveries can be achieved.