Learn More
Advanced Materials Technology HALO 120 A OLIGO C18 2.7 um 2.1 x 150 mm
Catalog No. NC3309651
Encompass_Preferred
The HALO OLIGO C18 incorporates surface modified organo-silane technology for alkaline resistance, resulting in excellent stability under elevated pH operating conditions common in oligonucleotide separation methods.  The inert column hardware also addresses absorptivity concerns so the highest recoveries can be achieved.
Catalog No. NC3309651 Supplier Advanced Materials Technology Supplier No. P2A62702
The HALO OLIGO C18 incorporates surface modified organo-silane technology for alkaline resistance, resulting in excellent stability under elevated pH operating conditions common in oligonucleotide separation methods.  The inert column hardware also addresses absorptivity concerns so the highest recoveries can be achieved.