Learn More
Advanced Materials Technology HALO 160 C30 2.7 10.0 X
Supplier:  Advanced Materials Technology 92110702
Encompass_Preferred
HALO C30 columns made with innovative Fused-Core particle technology for excellent reproducibility and column lifetimes provide fast, high resolution separations. Available in 2.7 µm particle size and a wide range of column dimensions, HALO 160 Å C30 columns are optimized for high performance in HPLC, UHPLC, and LCMS small molecule applications.
Catalog No. 50-255-6576
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.