Learn More
Advanced Materials Technology HALO 160 C30 2.7 4.6 X
Supplier: Advanced Materials Technology 92114930
HALO C30 columns made with innovative Fused-Core particle technology for excellent reproducibility and column lifetimes provide fast, high resolution separations. Available in 2.7 µm particle size and a wide range of column dimensions, HALO 160 Å C30 columns are optimized for high performance in HPLC, UHPLC, and LCMS small molecule applications.