Learn More
Advanced Materials Technology HALO 160 PFAS DELAY 2.7
Supplier:  Advanced Materials Technology 92114415
Encompass_Preferred
The HALO PFAS Delay column is optimized to prevent background PFAS contamination from interfering with the sample results. HALO PFAS Delay column is to be used with the HALO PFAS column.
Catalog No. 50-255-6524