Learn More
Advanced Materials Technology HALO 1000 Å Diphenyl, 2.7 µm, 2.1 x 50 mm
Catalog No. NC1665565
Encompass_Preferred
HALO BIOCLASS Diphenyl 1000 Å Protein columns made with innovative large pore Fused-Core particle technology provide best in class performance for high resolution separations and better recoveries of large biomolecules such as mAbs, versus smaller pore size columns. HALO 1000 Å columns are optimized for high performance in HPLC, UHPLC, and LCMS large molecule applications delivering excellent reproducibility and column lifetimes.
Catalog No. NC1665565 Supplier Advanced Materials Technology Supplier No. 92712426
HALO BIOCLASS Diphenyl 1000 Å Protein columns made with innovative large pore Fused-Core particle technology provide best in class performance for high resolution separations and better recoveries of large biomolecules such as mAbs, versus smaller pore size columns. HALO 1000 Å columns are optimized for high performance in HPLC, UHPLC, and LCMS large molecule applications delivering excellent reproducibility and column lifetimes.