Learn More
Advanced Materials Technology HALO 400 C4 3.4 1.0 X 1
Catalog No. 502556076
Encompass_Preferred
HALO BIOCLASS C4 400 Å Protein columns made with innovative large pore Fused-Core particle technology provide narrower peaks and better recoveries for high resolution separations of large biomolecules such as mAbs, versus smaller pore size columns. HALO 400 Å columns are optimized for high performance in HPLC, UHPLC, and LCMS large molecule applications delivering excellent reproducibility and column lifetimes.
Catalog No. 50-255-6076 Supplier Advanced Materials Technology Supplier No. 93411614
HALO BIOCLASS C4 400 Å Protein columns made with innovative large pore Fused-Core particle technology provide narrower peaks and better recoveries for high resolution separations of large biomolecules such as mAbs, versus smaller pore size columns. HALO 400 Å columns are optimized for high performance in HPLC, UHPLC, and LCMS large molecule applications delivering excellent reproducibility and column lifetimes.