Learn More
Advanced Materials Technology HALO 90 A C18 2.7UM 4.6X250MM
Catalog No. NC2468412
Encompass_Preferred
HALO C18 columns made with innovative Fused-Core particle technology for excellent reproducibility and column lifetimes provide fast, high resolution separations. Available in three particle sizes and a wide range of column dimensions, HALO 90 ¿C18 columns are optimized for high performance in HPLC, UHPLC, and LCMS small molecule applications.
Catalog No. NC2468412 Supplier Advanced Materials Technology Supplier No. 92814902
HALO C18 columns made with innovative Fused-Core particle technology for excellent reproducibility and column lifetimes provide fast, high resolution separations. Available in three particle sizes and a wide range of column dimensions, HALO 90 ¿C18 columns are optimized for high performance in HPLC, UHPLC, and LCMS small molecule applications.