Learn More
Advanced Materials Technology HALO 90 A GLYCAN 2.7 0.3
Catalog No. 502555578
Encompass_Preferred
HALO Glycan specific solution is ideal for HILIC separations. Offers high resolution even with complex glycan mixtures
Catalog No. 50-255-5578 Supplier Advanced Materials Technology Supplier No. 99226705
HALO Glycan specific solution is ideal for HILIC separations. Offers high resolution even with complex glycan mixtures