Learn More
Advanced Materials Technology HALO 90 A HILIC, 2.7, 2.1 x 50 mm - INERT
Catalog No. 502745702
Encompass_Preferred
HALO 90 A HILIC, 2.7, 2.1 x 50 mm - INERT
Catalog No. 50-274-5702 Supplier Advanced Materials Technology Supplier No. P2812401
HALO 90 A HILIC, 2.7, 2.1 x 50 mm - INERT