Learn More
Advanced Materials Technology HALO 90 AQ-C18 2.7 3.0
Catalog No. 502556231
Encompass_Preferred
HALO AQ-C18 columns made with innovative Fused-Core particle technology for excellent reproducibility and column lifetimes provide fast, high resolution separations. Available in three particle sizes and a wide range of column dimensions, HALO 90 Å AQ-C18 columns are optimized for high performance in HPLC, UHPLC, and LCMS small molecule applications.
Catalog No. 50-255-6231 Supplier Advanced Materials Technology Supplier No. 92813322
Add to Cart
Add to Cart
HALO AQ-C18 columns made with innovative Fused-Core particle technology for excellent reproducibility and column lifetimes provide fast, high resolution separations. Available in three particle sizes and a wide range of column dimensions, HALO 90 Å AQ-C18 columns are optimized for high performance in HPLC, UHPLC, and LCMS small molecule applications.