Learn More
Advanced Materials Technology HALO 90 AQ-C18 5 10.0 X
Catalog No. 502555918
Encompass_Preferred
HALO AQ-C18 columns made with innovative Fused-Core particle technology for excellent reproducibility and column lifetimes provide fast, high resolution separations. Available in three particle sizes and a wide range of column dimensions, HALO 90 Å AQ-C18 columns are optimized for high performance in HPLC, UHPLC, and LCMS small molecule applications.
Catalog No. 50-255-5918 Supplier Advanced Materials Technology Supplier No. 95810422
Add to Cart
Add to Cart
HALO AQ-C18 columns made with innovative Fused-Core particle technology for excellent reproducibility and column lifetimes provide fast, high resolution separations. Available in three particle sizes and a wide range of column dimensions, HALO 90 Å AQ-C18 columns are optimized for high performance in HPLC, UHPLC, and LCMS small molecule applications.