Learn More
Advanced Materials Technology HALO 90 AQ-C18 5 2.1 X
Supplier: Advanced Materials Technology 95812722
HALO AQ-C18 columns made with innovative Fused-Core particle technology for excellent reproducibility and column lifetimes provide fast, high resolution separations. Available in three particle sizes and a wide range of column dimensions, HALO 90 Å AQ-C18 columns are optimized for high performance in HPLC, UHPLC, and LCMS small molecule applications.