Learn More
Advanced Materials Technology HALO 90 BIPHENYL 2 3.0
Catalog No. 502556650
Encompass_Preferred
HALO Biphenyl columns made with innovative Fused-Core particle technology for excellent reproducibility and column lifetimes provide fast, high resolution separations. Available in three particle sizes and a wide range of column dimensions, HALO 90 Å Biphenyl columns are optimized for high performance in HPLC, UHPLC, and LCMS small molecule applications.
Catalog No. 50-255-6650 Supplier Advanced Materials Technology Supplier No. 91813111
HALO Biphenyl columns made with innovative Fused-Core particle technology for excellent reproducibility and column lifetimes provide fast, high resolution separations. Available in three particle sizes and a wide range of column dimensions, HALO 90 Å Biphenyl columns are optimized for high performance in HPLC, UHPLC, and LCMS small molecule applications.