Learn More
Advanced Materials Technology HALO 90 BIPHENYL 5 10.0
Catalog No. 502555919
Encompass_Preferred
HALO Biphenyl columns made with innovative Fused-Core particle technology for excellent reproducibility and column lifetimes provide fast, high resolution separations. Available in three particle sizes and a wide range of column dimensions, HALO 90 Å Biphenyl columns are optimized for high performance in HPLC, UHPLC, and LCMS small molecule applications.
Catalog No. 50-255-5919 Supplier Advanced Materials Technology Supplier No. 95810411
HALO Biphenyl columns made with innovative Fused-Core particle technology for excellent reproducibility and column lifetimes provide fast, high resolution separations. Available in three particle sizes and a wide range of column dimensions, HALO 90 Å Biphenyl columns are optimized for high performance in HPLC, UHPLC, and LCMS small molecule applications.