Learn More
Advanced Materials Technology HALO 90 BIPHENYL 5 4.6
Supplier: Advanced Materials Technology 95814211
HALO Biphenyl columns made with innovative Fused-Core particle technology for excellent reproducibility and column lifetimes provide fast, high resolution separations. Available in three particle sizes and a wide range of column dimensions, HALO 90 Å Biphenyl columns are optimized for high performance in HPLC, UHPLC, and LCMS small molecule applications.