Learn More
Advanced Materials Technology HALO 90 C18 5 4.6 X 50
Catalog No. 502555698
Encompass_Preferred
HALO C18 columns made with innovative Fused-Core particle technology for excellent reproducibility and column lifetimes provide fast, high resolution separations. Available in three particle sizes and a wide range of column dimensions, HALO 90 Å C18 columns are optimized for high performance in HPLC, UHPLC, and LCMS small molecule applications.
Catalog No. 50-255-5698 Supplier Advanced Materials Technology Supplier No. 95814402
HALO C18 columns made with innovative Fused-Core particle technology for excellent reproducibility and column lifetimes provide fast, high resolution separations. Available in three particle sizes and a wide range of column dimensions, HALO 90 Å C18 columns are optimized for high performance in HPLC, UHPLC, and LCMS small molecule applications.