Learn More
Advanced Materials Technology HALO 90 C8 2 2.1 X 75 M
Catalog No. 502556682
Encompass_Preferred
HALO C8 columns made with innovative Fused-Core particle technology for excellent reproducibility and column lifetimes provide fast, high resolution separations. Available in three particle sizes and a wide range of column dimensions, HALO 90 Å C8 columns are optimized for high performance in HPLC, UHPLC, and LCMS small molecule applications.
Catalog No. 50-255-6682 Supplier Advanced Materials Technology Supplier No. 91812508
HALO C8 columns made with innovative Fused-Core particle technology for excellent reproducibility and column lifetimes provide fast, high resolution separations. Available in three particle sizes and a wide range of column dimensions, HALO 90 Å C8 columns are optimized for high performance in HPLC, UHPLC, and LCMS small molecule applications.