Learn More
Advanced Materials Technology HALO 90 C8 2.7 10.0 X 1
Supplier: Advanced Materials Technology 92810608
HALO C8 columns made with innovative Fused-Core particle technology for excellent reproducibility and column lifetimes provide fast, high resolution separations. Available in three particle sizes and a wide range of column dimensions, HALO 90 Å C8 columns are optimized for high performance in HPLC, UHPLC, and LCMS small molecule applications.