Learn More
Advanced Materials Technology HALO 90 ES-CN 5 2.1 X 5
Catalog No. 502555856
Encompass_Preferred
HALO ES-CN columns made with innovative Fused-Core particle technology for excellent reproducibility and column lifetimes provide fast, high resolution separations. Available in three particle sizes and a wide range of column dimensions, HALO 90 Å ES-CN columns are optimized for high performance in HPLC, UHPLC, and LCMS small molecule applications.
Catalog No. 50-255-5856 Supplier Advanced Materials Technology Supplier No. 95812404
HALO ES-CN columns made with innovative Fused-Core particle technology for excellent reproducibility and column lifetimes provide fast, high resolution separations. Available in three particle sizes and a wide range of column dimensions, HALO 90 Å ES-CN columns are optimized for high performance in HPLC, UHPLC, and LCMS small molecule applications.