Learn More
Advanced Materials Technology HALO 90 GLYCAN 2.7 4.6
Catalog No. 502556091
Encompass_Preferred
HALO Glycan specific solution is ideal for HILIC separations. Offers high resolution even with complex glycan mixtures
Catalog No. 50-255-6091 Supplier Advanced Materials Technology Supplier No. 92924605
HALO Glycan specific solution is ideal for HILIC separations. Offers high resolution even with complex glycan mixtures