Learn More
Advanced Materials Technology HALO 90 HILIC 2.7 0.5 X
Supplier: Advanced Materials Technology 98215401
HALO HILIC columns made with innovative Fused-Core particle technology for excellent reproducibility and column lifetimes provide fast, high resolution separations. Available in three particle sizes and a wide range of column dimensions, HALO 90 Å HILIC columns are optimized for high performance in HPLC, UHPLC, and LCMS small molecule applications.