Learn More
Advanced Materials Technology HALO 90 HILIC 5 4.6 X 3
Catalog No. 502555709
Encompass_Preferred
HALO HILIC columns made with innovative Fused-Core particle technology for excellent reproducibility and column lifetimes provide fast, high resolution separations. Available in three particle sizes and a wide range of column dimensions, HALO 90 Å HILIC columns are optimized for high performance in HPLC, UHPLC, and LCMS small molecule applications.
Catalog No. 50-255-5709 Supplier Advanced Materials Technology Supplier No. 95814301
HALO HILIC columns made with innovative Fused-Core particle technology for excellent reproducibility and column lifetimes provide fast, high resolution separations. Available in three particle sizes and a wide range of column dimensions, HALO 90 Å HILIC columns are optimized for high performance in HPLC, UHPLC, and LCMS small molecule applications.