Learn More
Advanced Materials Technology HALO 90 LPH-C18 2.7 2.1
Supplier:  Advanced Materials Technology 92822116
Encompass_Preferred
HALO C18-LPH is a low pH compatible, 90 Å, superficially porous particle C18 phase useful for any chromatographer running under low pH conditions. The sterically protected ligand reduces acidic hydrolysis which enables low pH mobile phases to be used without sacrificing column performance over time.
Catalog No. 50-255-6126