Learn More
Advanced Materials Technology HALO 90 Å PCS C18, 2 µm, 3.0 mm x 50 mm
Catalog No. 5000332655
Encompass_Preferred
Built upon proven Fused-Core technology for speed and efficiency, the HALO PCS column products are positively charged surface chemistries designed to deliver improved peak shapes for basic compounds observed with standard Phenyl-Hexyl chemistry. Ideal for use with low ionic strength mobile phases, HALO PCS maintains peak symmetry at higher loading capacities and provides alternate selectivities from Phenyl-Hexyl bonded phases. The columns are optimized to deliver performance for reproducible, high efficiency LC and LCMS separations.
Catalog No. 50-003-32655 Supplier Advanced Materials Technology Supplier No. 91883417
Built upon proven Fused-Core technology for speed and efficiency, the HALO PCS column products are positively charged surface chemistries designed to deliver improved peak shapes for basic compounds observed with standard Phenyl-Hexyl chemistry. Ideal for use with low ionic strength mobile phases, HALO PCS maintains peak symmetry at higher loading capacities and provides alternate selectivities from Phenyl-Hexyl bonded phases. The columns are optimized to deliver performance for reproducible, high efficiency LC and LCMS separations.