Learn More
Advanced Materials Technology HALO 90 PENTA-HILIC 2 3
Supplier:  Advanced Materials Technology 91813305
Encompass_Preferred
HALO Penta-HILIC columns made with innovative Fused-Core particle technology for excellent reproducibility and column lifetimes provide fast, high resolution separations. Available in three particle sizes and a wide range of column dimensions, HALO 90 Å Penta-HILIC columns are optimized for high performance in HPLC, UHPLC, and LCMS small molecule applications.
Catalog No. 50-255-6636